4-Chloro-2-hydroxypyridine ≥98%

Supplier: Thermo Scientific
Warning

H29250.MD H29250.03
H29250.MDEA 85.4 EUR
H29250.MD H29250.03
4-Chloro-2-hydroxypyridine ≥98%
4-Chloro-2-hydroxypyridine

Purity: 98% CAS No.: 40673-25-4 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 177-179° MDL No.: MFCD03093022

Formula: C₅H₄ClNO
MW: 129,55 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD03093022
CAS Number: 40673-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR