Isoniazid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide
A10583.0B
A10583.18
A10583.22
A10583.36
A10583.0BEA
252
EUR
A10583.0B
A10583.18
A10583.22
A10583.36
Isoniazid ≥98%
Isoniazid
Purity: 98+%
CAS No.: 54-85-3
Molecular Formula: C6H7N3O
Formula Weight: 137.14
Melting Point: 169-174°
Beilstein Reference: 119374
Merck Reference: 14,5186
MDL No.: MFCD00006426
Formula:
C₆H₇N₃O MW: 137.14 g/mol Melting Pt: 169…174 °C Density: 1.42 g/cm³ (20 °C) Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006426 CAS Number: 54-85-3 EINECS: 200-214-6 Merck Index: 13,05205 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...