6-Hydroxynicotinic acid 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

156400250 156401000 156400050
ACRO156400250EA 72.8 EUR
ACRO156400250 ACRO156401000 ACRO156400050
6-Hydroxynicotinic acid 99%
6-Hydroxynicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006277
CAS Number: 5006-66-6
EINECS: 225-682-9

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR