2-Amino-5-iodopyridine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine
B25175.03
B25175.06
B25175.14
B25175.03EA
40.7
EUR
B25175.03
B25175.06
B25175.14
2-Amino-5-iodopyridine 97%
2-Amino-5-iodopyridine
Purity: 97%
CAS No.: 20511-12-0
Molecular Formula: C5H5IN2
Formula Weight: 220.01
Melting Point: 126-130°
Beilstein Reference: 108738
Formula:
C₅H₅IN₂ MW: 220.01 g/mol Boiling Pt: 271 Melting Pt: 126…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160312 CAS Number: 20511-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...