2-Amino-3-nitropyridine 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine
151120050
151120250
ACRO151120050EA
43.2
EUR
ACRO151120050
ACRO151120250
2-Amino-3-nitropyridine 99%
2-Amino-3-nitropyridine
Formula:
C₅H₅N₃O₂ Melting Pt: 165,9…166.4 °C Density: 1,54 g/cm³ (20 °C) Flash Pt: 167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006314 CAS Number: 4214-75-9 EINECS: 224-144-0 |
Specification Test Results
Appearance | Yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 164 to 168 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...