4-Amino-3-nitropyridine 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine
L19725.06
L19725.14
L19725.06EA
52.5
EUR
L19725.06
L19725.14
4-Amino-3-nitropyridine 96%
4-Amino-3-nitropyridine
Purity: 96%
CAS No.: 1681-37-4
Molecular Formula: C5H5N3O2
Formula Weight: 139.11
Melting Point: 201-205°
Beilstein Reference: 383710
Formula:
C₅H₅N₃O₂ MW: 139,11 g/mol Melting Pt: 201…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160304 CAS Number: 1681-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...