4-Pyridone 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
348830250
348831000
348835000
ACRO348830250EA
61.3
EUR
ACRO348830250
ACRO348831000
ACRO348835000
4-Pyridone 95%
4-Pyridone
Formula:
C₅H₅NO Boiling Pt: 230…235 °C (12 mmHg) Melting Pt: 140…147 °C Flash Pt: 221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006419 CAS Number: 626-64-2 EINECS: 210-958-3 |
Specification Test Results
Appearance | White to yellow to beige to light brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 139 to 151 °C |
HPLC | ≥94 % |
Titration with HClO4 | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...