3-Acetylpyridine ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone
A14246.14
A14246.22
A14246.36
A14246.14EA
49.1
EUR
A14246.14
A14246.22
A14246.36
3-Acetylpyridine ≥98%
3-Acetylpyridine
Purity: 98%
CAS No.: 350-03-8
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 104°(219°F)
Refractive Index: 1.5350
Odour: Characteristic
Melting Point: 12-13°
Boiling Point: 218-220°
Beilstein Reference: 107751
Merck Reference: 14,6116
UN No.: 2810
MDL No.: MFCD00006396
Formula:
C₇H₇NO MW: 121,14 g/mol Boiling Pt: 218…220 °C Melting Pt: 12…13 °C Density: 1,106 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006396 CAS Number: 350-03-8 EINECS: 206-496-7 UN: 2810 ADR: 6.1,II Merck Index: 14,6116 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...