2,4-Dichloropyrimidine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
162650250
162650050
162651000
ACRO162650250EA
170
EUR
ACRO162650250
ACRO162650050
ACRO162651000
2,4-Dichloropyrimidine 97%
2,4-Dichloropyrimidine
Formula:
C₄H₂Cl₂N₂ Boiling Pt: 101 °C (23 mmHg) Melting Pt: 57…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006061 CAS Number: 3934-20-1 EINECS: 223-508-6 |
Specification Test Results
Appearance | White to yellow to beige to grey Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 57 to 62 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...