2-Chloropyrimidine 98+%

Supplier: Thermo Scientific
Warning

185840100 185840500 185842500
ACRO185840100EA 47 EUR
ACRO185840100 ACRO185840500 ACRO185842500
2-Chloropyrimidine 98+%
2-Chloropyrimidine
Formula: C₄H₃ClN₂
MW: 114,53 g/mol
Boiling Pt: 75…76 °C (10 mmHg)
Melting Pt: 62…66 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to light orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR