3-Methoxythiophene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B21940.06
B21940.14
B21940.06EA
93.2
EUR
B21940.06
B21940.14
3-Methoxythiophene ≥99%
3-Methoxythiophene
Purity: 99%
CAS No.: 17573-92-1
Molecular Formula: C5H6OS
Formula Weight: 114.17
Flash Point: 49°(120°F)
Refractive Index: 1.5320
Odour: Recognizable
Boiling Point: 60-61°/12mm
Beilstein Reference: 106404
UN No.: 3271
MDL No.: MFCD00043894
Formula:
C₅H₆OS MW: 114.17 g/mol Boiling Pt: 60…61 °C (12 mmHg) Melting Pt: –7 °C Density: 1.147 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043894 CAS Number: 17573-92-1 UN: 3271 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...