3-Acetylthiophene ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Methyl 3-thienyl ketone, 1-(3-Thienyl)-1-ethanone

A11078.09 A11078.18 A11078.30
A11078.09EA 120 EUR
A11078.09 A11078.18 A11078.30
3-Acetylthiophene ≥98%
3-Acetylthiophene

Purity: 98% CAS No.: 1468-83-3 Molecular Formula: C6H6OS Formula Weight: 126.18 Storage Temperature: 2°C to 8°C Melting Point: 58-61° Boiling Point: 208-210° Beilstein Reference: 107241 UN No.: 2811 MDL No.: MFCD00005468

Formula: C₆H₆OS
MW: 126,18 g/mol
Boiling Pt: 208…210 °C
Melting Pt: 58…61 °C
MDL Number: MFCD00005468
CAS Number: 1468-83-3
EINECS: 215-996-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR