1,3,5-Trioxane 99+%
Supplier: Thermo Fisher Scientific
|
Danger
|
140290020
140295000
140290051
140291000
ACRO140290020EA
120
EUR
ACRO140290020
ACRO140295000
ACRO140290051
ACRO140291000
1,3,5-Trioxane 99+%
1,3,5-Trioxane
Formula:
C₃H₆O₃ Boiling Pt: 114…116 °C (760 mmHg) Melting Pt: 61…62 °C Density: 1,17 g/cm³ Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006563 CAS Number: 110-88-3 EINECS: 203-812-5 UN: 1325 ADR: 4.1,II Merck Index: 12,09863 |
Specification Test Results
Appearance | Colorless to white Crystals or crystalline flakes |
Infrared spectrum | Conforms |
Melting point | 60 to 63 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...