Advanced pharmaceutical refrigerators
Supplier: Labcold
RAFR44043 RAFG21043 RAFG44043 RAFR21043
BORORAFR44043EA 10070 EUR
BORORAFR44043 BORORAFG21043 BORORAFG44043 BORORAFR21043
Advanced pharmaceutical refrigerators
Refrigerators
Advanced ranges of refrigerators along with their pharmacy counterparts, represent the highest levels of performance, constructional quality and robustness.
  • Automatic defrosting
  • Adjustable shelves
  • Lockable doors
  • Microprocessor temperature control
  • Microprocessor control with ducted air flow and advanced auto defrost system provide class leading temperature stability and uniformity
  • High and low temperature alarms and min/max temperature recording
Heavy duty all-metal construction, with standard stainless steel interior provides strength and durability with ease of cleaning or sanitisation.
Temperature range: +2 to 8 °C.