ESD safety shoes, lace-up, Journey S1
Supplier: ELTEN GMBH
729630/40 729630/41 729630/42 729630/43 729630/44 729630/45 729630/46 729630/47 729630/48
ELTN729630/40EA 82.9 EUR
ELTN729630/40 ELTN729630/41 ELTN729630/42 ELTN729630/43 ELTN729630/44 ELTN729630/45 ELTN729630/46 ELTN729630/47 ELTN729630/48
ESD safety shoes, lace-up, Journey S1
Shoes and Boots
Sporty ESD safety shoes made from suede, mesh and microfibre upper, a skin friendly textile lining, a padded tongue plus a dual-density rubber/PU sole (E-Track).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole (Sportive ESD)
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, construction etc.
Colour: Grey
Certifications: EN 61340 (ESD); EN ISO 20345; S1.