Test and calibration station for portable gas detection instruments, X-dock®
Supplier: Dräger
8321891 8321892 8321901
DRAE8321891EA 1810 EUR
DRAE8321891 DRAE8321892 DRAE8321901
Test and calibration station for portable gas detection instruments, X-dock®
Docking Stations
The Dräger X-dock series provides full control of your portable gas detection instruments. Automatic bump tests and calibrations with reduced test gas consumption and short testing times save time and money. Comprehensive documentation and evaluations provide with a clear overview.
  • Immediately detects all sensor combinations
  • Short testing times reduce the overall maintenance efforts
  • Touchscreen operation at the master station
  • Up to three configurable test routines
  • Automatic leak tests and automatic tests of alarm elements (acoustic, visual and vibration alarms)
  • Replaceable seal cartridge with 12 V operation
  • Evaluation of sensor response time
The X-dock automatically tests and adjusts when all required test gases are connected. The station works independently and can be configured and used without a PC. The station is operated via an integrated touchscreen. A gas detection instrument is detected as soon as it is inserted, and all data is documented in the database.
The X-dock stores all of the relevant data and reads the collected information from the gas detection instruments for subsequent evaluations. The valve concept requires one pump only, resulting in the need for less wear and tear parts. The valves automatically switch the test gases and the compressed air.
Up to 10 modules can be connected to the X-dock at any given time, allowing to test gas detection instruments simultaneously. The automatic bump test also saves time, because the test only takes 8 to 15 seconds. All Dräger test gas cylinders are already stored in the database – entering the part number will automatically fill in the fields required for the gas configuration. This eliminates the need for manual data entry.