Laser labels, Tough-Spots®
Supplier: DISTRIBUTOR
SPOT1200 SPOT1300 SPOT1400 SPOT1500 SPOT2100 SPOT2200 SPOT2300 SPOT2400 SPOT2500 SPOT3000 SPOT4000
DIUKSPOT1200EA 0 EUR
DIUKSPOT1200 DIUKSPOT1300 DIUKSPOT1400 DIUKSPOT1500 DIUKSPOT2100 DIUKSPOT2200 DIUKSPOT2300 DIUKSPOT2400 DIUKSPOT2500 DIUKSPOT3000 DIUKSPOT4000
Laser labels, Tough-Spots®
Labels
Pre-cut, peel-off round labels for microcentrifuge tube tops. Chemically inert, polyolefin labels resist most organic solvents and caustic agents. Adhere to most plastics, glass, and metals without cracking, peeling, or degrading.
  • Specially top-coated to readily accept laser printing
  • Stay-flat sheets will not jam in laser printers
  • Withstand boiling water baths, moderate oven temperatures, autoclaving and extreme freezer temperatures
Fits a variety of microcentrifuge tube tops, cryovial caps and lids, laboratory container caps, lids and covers.