Sodium cyanide 95,0 ACS, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: 1-Dodecanesulphonic acid sodium salt

205222-100G 205222-2.5KG
205222-100GEA 0 EUR
205222-100G 205222-2.5KG
Sodium cyanide 95,0 ACS, Fluka™
Sodium cyanide
Formula: CNNa
MW: 49.01 g/mol
Boiling Pt: 1.407 °C (1.013 hPa)
Melting Pt: 563 °C
Density: 1.600 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1 I
Merck Index: 13,08679

Order Now

Specification Test Results

Assay Min. 95.0 %
Iron (Fe) Max. 0.005 %
Lead (Pb) Max. 5 ppm
Sulfide (S) Max. 0.005 %
Chloride (Cl) Max. 0.15 %
Phosphate (PO₄) Max. 0.02 %
Sulfate (SO₄) Max. 0.05 %
Thiocyanate (SCN) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR