Sodium hypophosphite monohydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
30410-50KG
30410-1KG
30410-50KGEA
3540
EUR
30410-50KG
30410-1KG
Sodium hypophosphite monohydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Sodium hypophosphite monohydrate
Formula:
NaH₂PO₂·H₂O Melting Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD06252379 CAS Number: 10039-56-2 EINECS: 231-669-9 |
Specification Test Results
Assay | Min. 99 % |
pH (5 %, 20°C) | 6-8 |
Ammonium (NH₄) | Max. 0.001 % |
Arsenic (As) | Max. 0.0001 % |
Barium (Ba) | Max. 0.005 % |
Calcium (Ca) | Max. 0.02 % |
Cadmium (Cd) | Max. 0.0005 % |
Cobalt (Co) | Max. 0.0005 % |
Copper (Cu) | Max. 0.0005 % |
Iron (Fe) | Max. 0.0005 % |
Magnesium (Mg) | Max. 0.005 % |
Nickel (Ni) | Max. 0.0005 % |
Lead (Pb) | Max. 0.0005 % |
Zinc (Zn) | Max. 0.0005 % |
Chloride (Cl) | Max. 0.005 % |
Phosphate, phosphite (as PO₄) | Max. 0.5 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...