1,4-Dioxane
Supplier: BDH US PRODUCED
|
Danger
|
BJ087-4
BJ087-1
BDHBBJ087-4EA
0
EUR
BDHBBJ087-4
BDHBBJ087-1
1,4-Dioxane
1,4-Dioxane
Formula:
C₄H₈O₂ Boiling Pt: 101,1 °C (1013 hPa) Melting Pt: 11,8 °C Density: 1,03 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006571 CAS Number: 123-91-1 EINECS: 204-661-8 UN: 1165 ADR: 3,II Merck Index: 13,03330 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...