Calcitriol ≥97%, crystalline solid

Supplier: Cayman Chemical
Danger

Synonyms: 1,25-Dihydroxycholecalciferol, (1R,3S,Z)-5-(2-((1R,3aS,7aR,E)-1-((R)-6-Hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha,25-Dihydroxyvitamin D3

71820-5 71820-10 71820-1
CAYM71820-5EA 504 EUR
CAYM71820-5 CAYM71820-10 CAYM71820-1
Calcitriol ≥97%, crystalline solid
Calcitriol

Calcitriol is synthesised from 7-dehydro cholesterol in humans by a non-enzymatic photochemical reaction with 290 to 310 nm UV light in the skin.


The main physiologic effects of calcitriol are to increase the absorption of calcium at the level of the intestinal epithelium, and to increase the mineralization of bone via the direct stimulation of osteoblasts.

Formula: C₂₇H₄₄O₃
MW: 416.646 g/mol
MDL Number: MFCD00867079
CAS Number: 32222-06-3
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Purity ≥97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR