Sinensetin ≥98%

Supplier: Cayman Chemical
Danger

Synonyms: 2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxy-4H-chromen-4-one, 2-(3,4-dimethoxyphenyl)-5,6,7-trimethoxy-1-benzopyran-4-one, Pedalitin permethyl ether, 2-(3,4-dimethoxyphenyl)-5,6,7-trimethoxychromen-4-one, 3`,4`,5,6,7-Pentamethoxyflavone

19751-10 19751-25 19751-5
CAYM19751-10EA 273 EUR
CAYM19751-10 CAYM19751-25 CAYM19751-5
Sinensetin ≥98%
Sinensetin

Sinensetin is a polymethoxylated flavone that is present in the Southeast Asian medical plant O. stamineus, as well as in orange oil. It has been shown to enhance adipogenesis and lipolysis by increasing cAMP levels in 3T3-L1 cells. Sinensetin is also reported to inhibit α-glucosidase and α-amylase activity in vitro, indicating its potential usefulness in the control of glucose absorption. Also it is used to suppress the expression of genes associated with inflammation by regulating IκBα protein levels in LPS-activated macrophages.

Formula: C₂₀H₂₀O₇
MW: 372,4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00017421
CAS Number: 2306-27-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity ≥98%
Formulation A Crystalline Solid
λmax 215, 241, 327 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR