4-Methyl-3-heptanol erythro + threo ≥99%

Supplier: Thermo Fisher Scientific
Warning

B20357.06 B20357.14
B20357.06EA 84.6 EUR
B20357.06 B20357.14
4-Methyl-3-heptanol erythro + threo ≥99%
4-Methyl-3-heptanol

Purity: 99% CAS No.: 14979-39-6 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 59°(138°F) Refractive Index: 1.4310 Boiling Point: 98-99°/75mm Beilstein Reference: 1733008 UN No.: 1987 MDL No.: MFCD00004568

Formula: C₈H₁₈O
Boiling Pt: 98…99 °C (75 mmHg)
Melting Pt: –123 °C
Density: 0,830
Flash Pt: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004568
CAS Number: 14979-39-6
EINECS: 239-058-9
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR