(±)-Ethyl-2-methylbutyrate ≥98%

Supplier: Thermo Fisher Scientific

L06743.AE L06743.AP
L06743.AEEA 35.8 EUR
L06743.AE L06743.AP
(±)-Ethyl-2-methylbutyrate ≥98%
(±)-Ethyl 2-methylbutyrate

Purity: 98% CAS No.: 7452-79-1 Molecular Formula: C7H14O2 Formula Weight: 130.19 Flash Point: 26°(78°F) Refractive Index: 1.3970 Boiling Point: 132-133° Beilstein Reference: 1720887 UN No.: 3272 MDL No.: MFCD00012217

Formula: C₇H₁₄O₂
MDL Number: MFCD00012217
CAS Number: 7452-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR