Hydrochloric acid 0.1 mol/l (0.1 N)

Supplier: SOUTHERN GROUP

Synonyms: Muriatic acidHydrochloric acid solution

SGRO0611EA 164 EUR
SGRO0611
Hydrochloric acid 0.1 mol/l (0.1 N)
Hydrochloric acid
Formula: HCl
MW: 36,46 g/mol
Density: 1,004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR