Hydrochloric acid 1 mol/l (1 N)

Supplier: SOUTHERN GROUP
Warning

Synonyms: Muriatic acidHydrochloric acid solution

SGRO0610EA 124 EUR
SGRO0610
Hydrochloric acid 1 mol/l (1 N)
Hydrochloric acid
Formula: HCl
MW: 36,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR