L(+)-Lysine monohydrochloride

Supplier: Roth Carl

Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl

9357.1 9357.2
ROTH9357.1EA 34.7 EUR
ROTH9357.1 ROTH9357.2
L(+)-Lysine monohydrochloride
L(+)-Lysine monohydrochloride
Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182,65 g/mol
Melting Pt: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2
EINECS: 211-519-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR