L(+)-Serine

Supplier: Roth Carl

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

4682.1 4682.2
ROTH4682.1EA 70.9 EUR
ROTH4682.1 ROTH4682.2
L(+)-Serine
L(+)-Serine
Formula: C₃H₇NO₃
MW: 105,09 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 215…225 °C
Density: 1,6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR