Lutetium trinitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Lutetium (III) nitrate hydrate
14568.03
14568.06
14568.14
14568.03EA
93.6
EUR
14568.03
14568.06
14568.14
Lutetium trinitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Lutetium trinitrate hydrate
Purity: 99.9%
CAS No.: 100641-16-5
Molecular Formula: LuN3O9.xH2O
Formula Weight: 360.98(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: White
UN No.: 1477
MDL No.: MFCD00150421
Formula:
LuN₃O₉·H₂O MW: 379 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150421 CAS Number: 100641-16-5 EINECS: 233-241-7 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...