Trichloroacetyl chloride ≥99%
Supplier: Thermo Scientific
|
Danger
|
B21675.0B
B21675.18
B21675.30
B21675.0BEA
669
EUR
B21675.0B
B21675.18
B21675.30
Trichloroacetyl chloride ≥99%
Trichloroacetyl chloride
Purity: 99%
CAS No.: 76-02-8
Molecular Formula: CCl3COCl
Formula Weight: 181.83
Refractive Index: 1.4700
Melting Point: -146°
Boiling Point: 114-116°
Beilstein Reference: 774120
UN No.: 2442
MDL No.: MFCD00000792
Formula:
C₂Cl₄O MW: 181,83 g/mol Boiling Pt: 114…116 °C Melting Pt: –146 °C Density: 1,629 Storage Temperature: Ambient |
MDL Number:
MFCD00000792 CAS Number: 76-02-8 EINECS: 200-926-7 UN: 2442 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...