Trichloroacetic anhydride 95%
Supplier: Thermo Scientific
|
Danger
|
L00849.18
L00849.30
L00849.18EA
92.1
EUR
L00849.18
L00849.30
Trichloroacetic anhydride 95%
Trichloroacetic anhydride
Purity: 95%
CAS No.: 4124-31-6
Molecular Formula: (Cl3CCO)2O
Formula Weight: 308.76
Refractive Index: 1.4840
Boiling Point: 98-100°/11mm
UN No.: 3265
MDL No.: MFCD00000793
Formula:
C₄Cl₆O₃ MW: 308,76 g/mol Boiling Pt: 98…100 °C (11 mmHg) Melting Pt: –2 °C Density: 1,690 Storage Temperature: Ambient |
MDL Number:
MFCD00000793 CAS Number: 4124-31-6 EINECS: 223-925-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...