Malononitrile ≥99%

Supplier: Thermo Scientific
Danger

A15046.22 A15046.06 A15046.0E A15046.36
A15046.22EA 31.2 EUR
A15046.22 A15046.06 A15046.0E A15046.36
Malononitrile ≥99%
Malononitrile

Purity: 99% CAS No.: 109-77-3 Molecular Formula: C3H2N2 Formula Weight: 66.06 Flash Point: 112°(233°F) Refractive Index: 1.4150 Melting Point: 30-34° Boiling Point: 220-222° Solubility: Soluble in water Beilstein Reference: 773697 Merck Reference: 14,5711 UN No.: 2647 MDL No.: MFCD00001883

Formula: C₃H₂N₂
MW: 66,06 g/mol
Boiling Pt: 220…222 °C
Melting Pt: 30…34 °C
Density: 1,049
Flash Pt: 112 °C (233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II
Merck Index: 14,5711

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR