2,3-Butanedione monoxime
Supplier: Roth Carl
Synonyms:
Diacetyl monoxime
3494.1
3494.2
ROTH3494.1EA
31.7
EUR
ROTH3494.1
ROTH3494.2
2,3-Butanedione monoxime
2,3-Butanedione monoxime
Formula:
C₄H₇NO₂ MW: 101,11 g/mol Boiling Pt: 185 °C (1013 hPa) Melting Pt: 74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002116 CAS Number: 57-71-6 EINECS: 200-348-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...