Trilaurin (glycerol trilaurate) 98%

Supplier: Thermo Scientific

Synonyms: Glyceryl tridodecanoate

421830250 421830050
ACRO421830250EA 609 EUR
ACRO421830250 ACRO421830050
Trilaurin (glycerol trilaurate) 98%
Trilaurin (Glycerol trilaurate)
Formula: C₃₉H₇₄O₆
MW: 639,01 g/mol
Boiling Pt: 643 °C (1013 hPa)
Melting Pt: 45…47 °C
Density: 0,9 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00026559
CAS Number: 538-24-9
EINECS: 208-687-0

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
TLC ≥97.5 %
pH 6.0 to 9.2 (5 % at 25°C)
Alkalinity ≤0.022 % (as KOH)
Iodate (IO3) ≤2 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Nitrogen compounds (as N) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR