Palmitoyl chloride 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
304291000
304295000
304290025
ACRO304291000EA
94
EUR
ACRO304291000
ACRO304295000
ACRO304290025
Palmitoyl chloride 98%
Palmitoyl chloride
Formula:
C₁₆H₃₁ClO Boiling Pt: 199 °C (2 mmHg) Melting Pt: 9…14 °C Density: 0,9 g/cm³ Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000742 CAS Number: 112-67-4 EINECS: 203-996-7 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4530 (20°C, 589 nm) |
Titration Argentometric | 98 to 101 % |
GC | ≥97.5 % |
Color scale | ≤80 APHA |
Acidity | ≤1 % (as palmitic acid) |
Phosphorus (P) | ≤150 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...