Stearyl stearate 98%
Supplier: Thermo Fisher Scientific
368110050
368110010
ACRO368110050EA
929
EUR
ACRO368110050
ACRO368110010
Stearyl stearate 98%
Stearyl stearate
Formula:
C₃₆H₇₂O₂ Boiling Pt: ∼175 °C (1013 hPa) Melting Pt: 64 °C Flash Pt: ≥ 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056225 CAS Number: 2778-96-3 EINECS: 220-476-5 |
Specification Test Results
Appearance | White Powder or platelets |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...