2-Butoxyethyl acetate for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Butylglycol acetate

M-Clarity™ quality level = MQ200

8.01395.1000 8.01395.2500
8.01395.1000EA 35.7 EUR
8.01395.1000 8.01395.2500
2-Butoxyethyl acetate for synthesis, Sigma-Aldrich®
2-Butoxyethyl acetate
Formula: C₈H₁₆O₃
MW: 160,21 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0,9416 g/cm³ (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00009457
CAS Number: 112-07-2
EINECS: 203-933-3

Order Now

Specification Test Results

Solubility 15 g/l (20 °C)
Melting Point -64 - -62 °C
Molar Mass 160.21 g/mol
Boiling Point 194 - 196 °C (1013 hPa)
Vapor Pressure 0.31 hPa (20 °C)
Flash Point 78 °C
Refractive Index 1.4138 (20 °C, 589 nm)
Explosion Limit 1.7 - 8.4 %(V)
Density 0.94 g/cm3 (20 °C)
pH 4 (10 g/l, H2O, 20 °C)
Ignition Point 280 °C
Evaporation number 190

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR