Ethyl iodoacetate 98% stabilised

Supplier: Thermo Fisher Scientific
Danger

222190100 222190500 222192500
ACRO222190100EA 63.4 EUR
ACRO222190100 ACRO222190500 ACRO222192500
Ethyl iodoacetate 98% stabilised
Ethyl iodoacetate
Formula: C₄H₇IO₂
MW: 214 g/mol
Boiling Pt: 179…180 °C (760 mmHg)
Density: 1.8 g/cm³
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001081
CAS Number: 623-48-3
EINECS: 210-796-3
UN: 2810
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear light yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5025 to 1.5055 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR