1-Pentanesulphonic acid sodium salt
Supplier: Molekula
Synonyms:
Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate
15583676-100G
15583676-25G
MOLE15583676-100GEA
263
EUR
MOLE15583676-100G
MOLE15583676-25G
1-Pentanesulphonic acid sodium salt
1-Pentanesulphonic acid sodium salt
Formula:
C₅H₁₁O₃SNa MW: 174,2 g/mol Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 EINECS: 245-208-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...