Dimethyl itaconate 97% stabilised
Supplier: Thermo Fisher Scientific
149161000
149165000
149160025
ACRO149161000EA
47.3
EUR
ACRO149161000
ACRO149165000
ACRO149160025
Dimethyl itaconate 97% stabilised
Dimethyl itaconate
Formula:
C₇H₁₀O₄ Boiling Pt: 208 °C (760 mmHg) Melting Pt: 34…39 °C Density: 1,24 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008446 CAS Number: 617-52-7 EINECS: 210-519-6 |
Specification Test Results
Appearance | White to light yellow Crystalline mass or liquid |
Infrared spectrum | Conforms |
Melting point | 34 to 39 °C |
GC | ≥96.0 % |
Stabilizer | 100 ppm hydroquinone |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...