2,2,4,4-Tetramethylpentane ≥98%
Supplier: Thermo Scientific
|
Danger
|
B21642.09
B21642.18
B21642.09EA
102
EUR
B21642.09
B21642.18
2,2,4,4-Tetramethylpentane ≥98%
2,2,4,4-Tetramethylpentane
Purity: 98%
CAS No.: 1070-87-7
Molecular Formula: C9H20
Formula Weight: 128.26
Flash Point: 6°(43°F)
Odour: Petroleum-like
Boiling Point: 121°
UN No.: 3295
MDL No.: MFCD00060873
Formula:
C₉H₂₀ MW: 128,26 g/mol Boiling Pt: 121 °C Melting Pt: –66,5 °C Density: 0,72 Flash Pt: 6 °C (43 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060873 CAS Number: 1070-87-7 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...