Penicillin G sodium salt

Supplier: Molekula

Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

47080913-25G 47080913-5G
MOLE47080913-25GEA 113 EUR
MOLE47080913-25G MOLE47080913-5G
Penicillin G sodium salt
Penicillin G sodium salt
Formula: C₁₆H₁₇N₂NaO₄S
MW: 356,38 g/mol
Melting Pt: 255 °C
Density: 1,38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00069666
CAS Number: 69-57-8
EINECS: 200-710-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR