3-Chloro-2-fluoroaniline ≥97%

Supplier: Thermo Scientific
Warning

H51144.06 H51144.22
H51144.06EA 33.5 EUR
H51144.06 H51144.22
3-Chloro-2-fluoroaniline ≥97%
3-Chloro-2-fluoroaniline

Purity: 97% CAS No.: 2106-04-9 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Flash Point: 110°(230°F) Refractive Index: 1.564 Odour: Characteristic Boiling Point: 214° UN No.: 2810 MDL No.: MFCD00069415

Formula: C₆H₅ClFN
MW: 145,56 g/mol
Boiling Pt: 214 °C
Density: 1,324
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00069415
CAS Number: 2106-04-9
EINECS: 218-283-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR