2-Chloro-4-nitrophenyl isocyanate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Chloro-2-isocyanato-5-nitrobenzene
L12685.03
L12685.06
L12685.03EA
29.9
EUR
L12685.03
L12685.06
2-Chloro-4-nitrophenyl isocyanate ≥97%
2-Chloro-4-nitrophenyl isocyanate
Purity: 97%
CAS No.: 40397-95-3
Molecular Formula: C7H3ClN2O3
Formula Weight: 198.57
Storage Temperature: 2°C to 8°C
Melting Point: 59-63°
Beilstein Reference: 2841619
UN No.: 2811
MDL No.: MFCD00037063
Formula:
C₇H₃ClN₂O₃ MW: 198,57 g/mol Melting Pt: 59…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037063 CAS Number: 40397-95-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...