2-Chloro-4-methoxyphenol ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Chlor-4-hydroxyanisole
A16652.06
A16652.14
A16652.06EA
55.1
EUR
A16652.06
A16652.14
2-Chloro-4-methoxyphenol ≥97%
2-Chloro-4-methoxyphenol
Purity: 97%
CAS No.: 18113-03-6
Molecular Formula: C7H7ClO2
Formula Weight: 158.59
Flash Point: 85°(185°F)
Odour: Phenol-like
Beilstein Reference: 1936412
MDL No.: MFCD00070773
Formula:
C₇H₇ClO₂ MW: 158,58 g/mol Boiling Pt: 242 °C (1013 hPa) Melting Pt: 39…44 °C Density: 1,153 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070773 CAS Number: 18113-03-6 EINECS: 242-007-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...