Ethyl-3,4,5-trihydroxybenzoate 99%
Supplier: Thermo Scientific
Synonyms:
Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate
279931000
279935000
ACRO279931000EA
78.3
EUR
ACRO279931000
ACRO279935000
Ethyl-3,4,5-trihydroxybenzoate 99%
Ethyl 3,4,5-trihydroxybenzoate
Formula:
C₉H₁₀O₅ MW: 198,18 g/mol Melting Pt: 151…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016430 CAS Number: 831-61-8 EINECS: 212-608-5 |
Specification Test Results
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 151 to 154 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...