Barium diphenylamine-4-sulphonate, Supelco®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
D4130-25G
D4130-50G
D4130-5G
D4130-25GEA
298
EUR
D4130-25G
D4130-50G
D4130-5G
Barium diphenylamine-4-sulphonate, Supelco®
Barium diphenylamine-4-sulphonate
Formula:
C₂₄H₂₀BaN₂O₆S₂ Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007497 CAS Number: 6211-24-1 EINECS: 228-278-0 UN: 1564 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...