Benzeneselenenyl bromide ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phenylselenyl bromide, Phenyl hypobromoselenoite, Bromoselenobenzene, Phenylselenenyl bromide

A11906.03 A11906.06 A11906.14
A11906.03EA 26 EUR
A11906.03 A11906.06 A11906.14
Benzeneselenenyl bromide ≥98%
Benzeneselenenyl bromide

Purity: 98% CAS No.: 34837-55-3 Molecular Formula: C6H5BrSe Formula Weight: 235.98 Melting Point: 56-62° Boiling Point: 107-108°/15mm UN No.: 2928 MDL No.: MFCD00000047

Formula: (C₆H₅)SeBr
Boiling Pt: 107…108 °C (15 mmHg)
Melting Pt: 56…62 °C
Density: 1,898 g/cm³ (65.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000047
CAS Number: 34837-55-3
EINECS: 252-238-1
UN: 2928
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR