3-Chloro-4-iodobenzotrifluoride ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14685.03
A14685.06
A14685.03EA
0
EUR
A14685.03
A14685.06
3-Chloro-4-iodobenzotrifluoride ≥98%
3-Chloro-4-iodobenzotrifluoride
Purity: 98%
CAS No.: 141738-80-9
Molecular Formula: C7H3ClF3I
Formula Weight: 306.45
Boiling Point: 58-60°/0.2mm
MDL No.: MFCD00042562
Formula:
C₇H₃ClF₃I Boiling Pt: 58…60 °C (0,2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00042562 CAS Number: 141738-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...