4-Nitro-o-toluic acid ≥97%

Supplier: Thermo Scientific
Warning

H31332.03 H31332.06
H31332.03EA 62.6 EUR
H31332.03 H31332.06
4-Nitro-o-toluic acid ≥97%
4-Nitro-o-toluic acid

Purity: 97% CAS No.: 1975-51-5 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Odour: Odourless Melting Point: 152-155° MDL No.: MFCD00210697

Formula: C₈H₇NO₄
MW: 181,15 g/mol
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00210697
CAS Number: 1975-51-5
EINECS: 217-828-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR